Terbium, Plasma Standard Solution, Specpure®, Tb 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
13860-AE
13860-AP
CAAA13860-AEEA
141.41
CAD
CAAA13860-AE
CAAA13860-AP
Terbium, Plasma Standard Solution, Specpure®, Tb 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Terbium Standards
MDL: MFCD00011256
Molecular Formula: Tb4O7 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011256
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...