Neodymium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

Synonyms: Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide

13837-14 13837-22
CAAA13837-14EA 210.14 CAD
CAAA13837-14 CAAA13837-22
Neodymium(III) oxide ≥99.997% (REO, rare earth oxide basis), REacton®
Neodymium (III) oxide

MDL: MFCD00011134


Purity: 99.997% CAS No.: 1313-97-9 Molecular Formula: Nd2O3 Formula Weight: 336.48 Physical Form: Powder Density: 7.24 Melting Point: 2272° Merck Reference: 14,6450 MDL No.: MFCD00011134

Formula: Nd₂O₃
Boiling Pt: 3760 °C (1013 hPa)
Melting Pt: 2272 °C
Density: 7.24
Storage Temperature: Ambient
MDL Number: MFCD00011134
CAS Number: 1313-97-9
EINECS: 215-214-1
Merck Index: 14,6450

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR