Potassium hydrogen sulfate mixture of potassium pyrosulphate, K2S2O7, and potassium bisulphate, KHSO4 ACS, fused

Supplier: Thermo Scientific Chemicals
Danger

13800-22 13800-36
CAAA13800-22EA 100.17 CAD
CAAA13800-22 CAAA13800-36
Potassium hydrogen sulfate mixture of potassium pyrosulphate, K2S2O7, and potassium bisulphate, KHSO4 ACS, fused
Potassium hydrogen sulphate

MDL: MFCD00066385


Formula Weight: 136.17 Physical Form: Fused Powder Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 214° UN No.: 2509 MDL No.: MFCD00066385

Formula: KHSO₄
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now

Specification Test Results

Acidity as H2SO3 37.5-38.6%
H2O 0.025
Cl 0.002%
PO3 0.00001
Ca 0.002%
MG 0.001%
Heavy metals (as Pb) 0.001%
Fe 0.002%
NA 0.01%%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR