Chromium ≥99.95% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

13796-14 13796-22 13796-36
CAAA13796-14EA 154.52 CAD
CAAA13796-14 CAAA13796-22 CAAA13796-36
Chromium ≥99.95% (metals basis), powder -200 mesh
Chromium

MDL: MFCD00010944


Purity: 99.95% CAS No.: 7440-47-3 Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00010944

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1 III
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR