Indium(III) acetylacetonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13794-03
13794-06
13794-14
CAAA13794-03EA
53.88
CAD
CAAA13794-03
CAAA13794-06
CAAA13794-14
Indium(III) acetylacetonate 98%
Indium (III) acetylacetonate
Used in catalysts
Purity: 98%
CAS No.: 14405-45-9
Molecular Formula: C15H21InO6
Formula Weight: 412.15
Physical Form: Powder
Melting Point: 186° dec.
MDL No.: MFCD00013494
Used in catalysts
Formula:
C₁₅H₂₁InO₆ Melting Pt: 186 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013494 CAS Number: 14405-45-9 EINECS: 238-378-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...