Vanadium ≥99.8% (metals basis), foil, Thickness: 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

13783-FY 13783-GK
CAAA13783-FYEA 246.48 CAD
CAAA13783-FY CAAA13783-GK
Vanadium ≥99.8% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Vanadium

MDL: MFCD00011453


Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈3.87g/50x100mm Odor: Odorless MDL No.: MFCD00011453

Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477 °C (1013 hPa)
Melting Pt: 1896 °C
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR