Hexaamminecobalt trichloride ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

13769-14 13769-22
CAAA13769-14EA 170.64 CAD
CAAA13769-14 CAAA13769-22
Hexaamminecobalt trichloride ≥97%
Hexaamminecobalt trichloride

MDL: MFCD00036304


Purity: 97% CAS No.: 10534-89-1 Molecular Formula: [Co(NH3)6]Cl3 Formula Weight: 267.48 Physical Form: Crystalline Density: 1.71 Merck Reference: 14,4675 MDL No.: MFCD00036304

Formula: H₁₈CoCl₃N₆
MW: 267.47 g/mol
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00036304
CAS Number: 10534-89-1
EINECS: 234-103-9
Merck Index: 14,4675

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR