Scandium(III) iodide, ultra dry ≥99.999% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

13687-03 13687-06
CAAA13687-03EA 462.94 CAD
CAAA13687-03 CAAA13687-06
Scandium(III) iodide, ultra dry ≥99.999% (REO, rare earth oxide basis)
Scandium (III) iodide

MDL: MFCD00054037


Purity: 99.999% CAS No.: 14474-33-0 Molecular Formula: ScI3 Formula Weight: 425.67 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00054037

Formula: I₃Sc
MW: 425.67
Melting Pt: 945 °C
Storage Temperature: Ambient
MDL Number: MFCD00054037
CAS Number: 14474-33-0
EINECS: 238-469-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR