Potassium chloride, dry_ultra 99,998 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassiumchloride

13682-09 13682-18
CAAA13682-09EA 203.82 CAD
CAAA13682-09 CAAA13682-18
Potassium chloride, dry_ultra 99,998 Metals basis
Potassium chloride

MDL: MFCD00011360 Fieser: 6,480


Purity: 99.998% CAS No.: 7447-40-7 Molecular Formula: KCl Formula Weight: 74.55 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 1.984 Refractive Index: 1.490 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 773° Boiling Point: 1500° subl. Merck Reference: 14,7621 MDL No.: MFCD00011360

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420 (subl.)
Melting Pt: 773
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR