Terbium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
13672-03
13672-06
CAAA13672-03EA
142.36
CAD
CAAA13672-03
CAAA13672-06
Terbium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Terbium (III) iodide
MDL: MFCD00049573
Fieser: 5,327 6,227
Purity: 99.99%
CAS No.: 13813-40-6
Molecular Formula: TbI3
Formula Weight: 539.64
Physical Form: Flake, Ampouled under argon
Sensitivity: Hygroscopic
MDL No.: MFCD00049573
Formula:
TbI₃ Melting Pt: 946 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049573 CAS Number: 13813-40-6 EINECS: 237-472-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...