Cadmium(II) chloride, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cadmium chloride
13667-06
13667-14
CAAA13667-06EA
150.73
CAD
CAAA13667-06
CAAA13667-14
Cadmium(II) chloride, ultra dry ≥99.998% (metals basis)
Cadmium (II) chloride
MDL: MFCD00010916
Purity: 99.998%
CAS No.: 10108-64-2
Molecular Formula: CdCl2
Formula Weight: 183.32
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.047
Melting Point: 568°
Boiling Point: 960°
Merck Reference: 14,1617
UN No.: 2570
MDL No.: MFCD00010916
Formula:
CdCl₂ MW: 183.32 g/mol Boiling Pt: 967 Melting Pt: 565 Density: 4.047 Storage Temperature: Ambient |
MDL Number:
MFCD00010916 CAS Number: 10108-64-2 EINECS: 233-296-7 UN: 2570 ADR: 6.1 III Merck Index: 14,1617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...