Cobalt(II) chloride, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cobaltous chloride, Cobalt dichloride
13665-03
13665-06
CAAA13665-03EA
95.75
CAD
CAAA13665-03
CAAA13665-06
Cobalt(II) chloride, ultra dry ≥99.998% (metals basis)
Cobalt (II) chloride
MDL: MFCD00010938
Decomposes at 400°C on long heating in air
Purity: 99.998%
CAS No.: 7646-79-9
Molecular Formula: Cl2Co
Formula Weight: 129.84
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.367
Appearance: Pale blue
Odor: Odorless
Melting Point: 735°
Boiling Point: 1049°
Merck Reference: 14,2437
UN No.: 3077
MDL No.: MFCD00010938
Formula:
Cl₂Co Boiling Pt: 1049 °C Melting Pt: 735 °C Density: 3.367 Storage Temperature: Ambient |
MDL Number:
MFCD00010938 CAS Number: 7646-79-9 EINECS: 231-589-4 UN: 3077 ADR: 9,III Merck Index: 14,2437 |
Specification Test Results
Form | -10 Mesh Beads, Ampouled under argon |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...