Neodymium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13661-14 13661-22
CAAA13661-14EA 199.08 CAD
CAAA13661-14 CAAA13661-22
Neodymium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium trifluoride

MDL: MFCD00011135


Purity: 99.9% CAS No.: 13709-42-7 Molecular Formula: NdF3 Formula Weight: 201.24 Physical Form: Crystalline Lump Density: 6.5 Appearance: Pale purple Odor: Odorless Melting Point: 1410° MDL No.: MFCD00011135

Formula: F₃Nd
MW: 201.24
Boiling Pt: 2327
Melting Pt: 1410
Density: 6.5
Storage Temperature: Ambient
MDL Number: MFCD00011135
CAS Number: 13709-42-7
EINECS: 237-253-3
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR