Samarium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

13660-14 13660-22
CAAA13660-14EA 298.62 CAD
CAAA13660-14 CAAA13660-22
Samarium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Samarium (III) fluoride

MDL: MFCD00049540


Purity: 99.9% CAS No.: 13765-24-7 Molecular Formula: SmF3 Formula Weight: 207.35 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049540

Formula: F₃Sm
MW: 207.36
Boiling Pt: 2323
Melting Pt: 1306 °C
Density: 4.362 g/cm³ (2327 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049540
CAS Number: 13765-24-7
EINECS: 237-367-3
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR