Europium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
13659-04
13659-09
CAAA13659-04EA
151.36
CAD
CAAA13659-04
CAAA13659-09
Europium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Europium (III) fluoride
MDL: MFCD00010998
Purity: 99.9%
CAS No.: 13765-25-8
Molecular Formula: EuF3
Formula Weight: 208.96
Physical Form: Crystalline
Sensitivity: Hygroscopic
Boiling Point:
Formula:
EuF₃ Boiling Pt: >2280 °C Melting Pt: 1390 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010998 CAS Number: 13765-25-8 EINECS: 237-368-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...