Terbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13657-06 13657-14
CAAA13657-06EA 199.08 CAD
CAAA13657-06 CAAA13657-14
Terbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Terbium (III) fluoride

MDL: MFCD00049572


Purity: 99.9% CAS No.: 13708-63-9 Molecular Formula: TbF3 Formula Weight: 215.92 Physical Form: Crystalline Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049572

Formula: TbF₃
Boiling Pt: 2277 °C (1013 hPa)
Melting Pt: 1172 °C
Density: 7.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049572
CAS Number: 13708-63-9
EINECS: 237-247-0
UN: 3288
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR