Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13656-14 13656-22
CAAA13656-14EA 149.94 CAD
CAAA13656-14 CAAA13656-22
Dysprosium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Dysprosium trifluoride

MDL: MFCD00016071


Purity: 99.9% CAS No.: 13569-80-7 Molecular Formula: DyF3 Formula Weight: 219.50 Physical Form: Crystalline Lump Density: 7.5 Appearance: White Odor: Odorless Melting Point: 1360° Boiling Point: 2200° MDL No.: MFCD00016071

Formula: DyF₃
Boiling Pt: 2200 °C
Melting Pt: 1360 °C
Density: 7.5
Storage Temperature: Ambient
MDL Number: MFCD00016071
CAS Number: 13569-80-7
EINECS: 236-992-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR