Gadolinium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

13655-14 13655-22
CAAA13655-14EA 173.8 CAD
CAAA13655-14 CAAA13655-22
Gadolinium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Gadolinium trifluoride

MDL: MFCD00016113


Purity: 99.9% CAS No.: 13765-26-9 Molecular Formula: GdF3 Formula Weight: 214.25 Physical Form: Crystalline Density: 7.1 Appearance: White Odor: Odorless Melting Point: 1231° MDL No.: MFCD00016113

Formula: F₃Gd
Melting Pt: 1231 °C
Density: 7.1
Storage Temperature: Ambient
MDL Number: MFCD00016113
CAS Number: 13765-26-9
EINECS: 237-369-4
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR