Yttrium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

13650-14 13650-36
CAAA13650-14EA 108.23 CAD
CAAA13650-14 CAAA13650-36
Yttrium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Yttrium (III) fluoride

MDL: MFCD00049714


Purity: 99.9% CAS No.: 13709-49-4 Molecular Formula: YF3 Formula Weight: 145.90 Physical Form: Lump/powder Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049714

Formula: F₃Y
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1387 °C
Density: 4.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049714
CAS Number: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR