Cerium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cerium (III) iodide, Cerous iodide
13641-03
13641-06
CAAA13641-03EA
145.52
CAD
CAAA13641-03
CAAA13641-06
Cerium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Cerium triiodide
MDL: MFCD00049423
Notes: Decomposes in moist air
Soluble in water
Purity: 99.9%
CAS No.: 7790-87-6
Molecular Formula: CeI3
Formula Weight: 520.83
Physical Form: Powder, Ampouled under argon
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Yellow
Odor: Odorless
Melting Point: 766°
Solubility: Soluble in water
Merck Reference: 14,1999
MDL No.: MFCD00049423
Formula:
CeI₃ MW: 520.83 g/mol Boiling Pt: 1400 °C (1013 hPa) Melting Pt: 766 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049423 CAS Number: 7790-87-6 EINECS: 232-228-3 Merck Index: 14,1999 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...