Holmium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

13635-03 13635-06
CAAA13635-03EA 172.22 CAD
CAAA13635-03 CAAA13635-06 CAAA13635-14
Holmium triiodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Holmium triiodide

MDL: MFCD00049745


Purity: 99.99% CAS No.: 13813-41-7 Molecular Formula: HoI3 Formula Weight: 545.64 Physical Form: Flakes, Ampouled under argon Sensitivity: Hygroscopic Merck Reference: 14,4727 MDL No.: MFCD00049745

Formula: HoI₃
MW: 545.64
Storage Temperature: Ambient
MDL Number: MFCD00049745
CAS Number: 13813-41-7
EINECS: 237-470-3
Merck Index: 14,4727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR