Tin(IV) iodide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
13617-06
13617-14
CAAA13617-06EA
287.56
CAD
CAAA13617-06
CAAA13617-14
Tin(IV) iodide, ultra dry ≥99.998% (metals basis)
Tin (IV) iodide
MDL: MFCD00036272
Purity: 99.998%
CAS No.: 7790-47-8
Molecular Formula: SnI4
Formula Weight: 626.31
Physical Form: Powder, Ampouled under argon
Density: 4.46
Refractive Index: 2.106
Sensitivity: Moisture Sensitive
Melting Point: ca 143°
Boiling Point: 340°
Merck Reference: 14,8777
UN No.: 3260
MDL No.: MFCD00036272
Formula:
I₄Sn Boiling Pt: 340 °C Melting Pt: ∼ 143 °C Density: 4.46 Storage Temperature: Ambient |
MDL Number:
MFCD00036272 CAS Number: 7790-47-8 EINECS: 232-208-4 UN: 3260 ADR: 8,III Merck Index: 14,8777 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...