Lithium iodide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
13600-06
13600-14
13600-22
CAAA13600-06EA
167.48
CAD
CAAA13600-06
CAAA13600-14
CAAA13600-22
Lithium iodide, ultra dry ≥99.999% (metals basis)
Lithium iodide
MDL: MFCD00011092
Purity: 99.999%
CAS No.: 10377-51-2
Molecular Formula: LiI
Formula Weight: 133.84
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.076
Refractive Index: 1.955
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White to off-white
Odor: Odorless
Melting Point: 449°
Boiling Point: 1170-1190°
Merck Reference: 14,5535
MDL No.: MFCD00011092
Formula:
ILi Boiling Pt: 1170…1190 °C Melting Pt: 449 °C Density: 4.076 Storage Temperature: Ambient |
MDL Number:
MFCD00011092 CAS Number: 10377-51-2 EINECS: 233-822-5 Merck Index: 14,5535 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...