Terbium trinitrate hydrate 99.99% (REO, rare earth oxide basis), crystalline aggregates, REacton®

Supplier: Thermo Scientific Chemicals
Danger

13593-04 13593-09 13593-18
CAAA13593-04EA 110.76 CAD
CAAA13593-04 CAAA13593-09 CAAA13593-18
Terbium trinitrate hydrate 99.99% (REO, rare earth oxide basis), crystalline aggregates, REacton®
Terbium trinitrate hydrate

Terbium(III) nitrate hydrate is used in ceramics, glass, phosphors, lasers, and also is the important dopant for fibre amplifiers. It is also used in alloys and in the production of electronic devices.


Purity: 99.99% Molecular Formula: Tb(NO3)3.xH2O Formula Weight: 344.94(anhy) Physical Form: Crystalline Aggregates, Packaged under argon Merck Reference: 14,9157 UN No.: 1477 MDL No.: MFCD00011257


Caution: Store away from metal powders and reducing agents. Incompatible with organic materials.

Formula: N₃O₉Tb·xH₂O
Melting Pt: 89.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00011257
CAS Number: 10043-27-3
EINECS: 233-138-7
UN: 1477
ADR: 5.1,III
Merck Index: 14,9157

Order Now

Specification Test Results

Total Rare Earth Oxide Impurities 0.01% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR