Rubidium iodide ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
13497-09
13497-18
CAAA13497-09EA
113.92
CAD
CAAA13497-09
CAAA13497-18
Rubidium iodide ≥99.8% (metals basis)
Rubidium iodide
MDL: MFCD00011192
Freely soluble in water. Soluble in alcohol
Purity: 99.8%
CAS No.: 7790-29-6
Molecular Formula: RbI
Formula Weight: 212.37
Physical Form: Crystalline
Density: 3.55
Refractive Index: 1.6474
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 647°
Boiling Point: 1300°
Solubility: Freely soluble in water. Soluble in alcohol
Merck Reference: 14,8286
MDL No.: MFCD00011192
Formula:
IRb Boiling Pt: 1300 °C Melting Pt: 647 °C Density: 3.55 Storage Temperature: Ambient |
MDL Number:
MFCD00011192 CAS Number: 7790-29-6 EINECS: 232-198-1 Merck Index: 14,8286 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...