Rubidium sulfate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
13493-36
13493-14
13493-22
CAAAA13493-36EA
2363.68
CAD
CAAAA13493-36
CAAA13493-14
CAAA13493-22
Rubidium sulfate ≥99% (metals basis)
Rubidium sulphate
MDL: MFCD00011190
Purity: 99%
CAS No.: 7488-54-2
Molecular Formula: Rb2SO4
Formula Weight: 267.00
Physical Form: Crystalline
Density: 3.613
Refractive Index: 1.513
Odor: Odorless
Melting Point: 1050°
MDL No.: MFCD00011190
Formula:
O₄Rb₂S Boiling Pt: 1708 °C (1013 hPa) Melting Pt: 1050 °C Density: 3,613 Storage Temperature: Ambient |
MDL Number:
MFCD00011190 CAS Number: 7488-54-2 EINECS: 231-301-7 |
Specification Test Results
Form | Crystalline |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...