Cesium chromate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

13492-14 13492-22
CAAA13492-14EA 121.66 CAD
CAAA13492-14 CAAA13492-22
Cesium chromate ≥99.9% (metals basis)
Cesium chromate

MDL: MFCD00016050


Purity: 99.9% CAS No.: 13454-78-9 Molecular Formula: Cs2CrO4 Formula Weight: 381.81 Physical Form: Powder Sensitivity: Hygroscopic UN No.: 3087 MDL No.: MFCD00016050

Formula: CrCs₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00016050
CAS Number: 13454-78-9
EINECS: 236-640-4
UN: 3087
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR