Cesium bromide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Crystals for infrared spectroscopy, x-ray fluorescent screens, scintillation counters
Purity: 99%
CAS No.: 7787-69-1
Molecular Formula: BrCs
Formula Weight: 212.81
Physical Form: Crystalline
Density: 4.44
Refractive Index: 1.6984
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 636°
Boiling Point: 1300°
Solubility: Very soluble in water. Soluble in alcohol. Insoluble in acetone
Merck Reference: 14,2009
MDL No.: MFCD00010954
Crystals for infrared spectroscopy, x-ray fluorescent screens, scintillation counters
Formula:
BrCs Boiling Pt: 1300 °C Melting Pt: 636 °C Density: 4.44 Storage Temperature: Ambient |
MDL Number:
MFCD00010954 CAS Number: 7787-69-1 EINECS: 232-130-0 Merck Index: 14,2009 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...