Copper ≥99.9% (metals basis), tubing 3.175 mm (0.125 in) OD, 1.5 mm (0.059 in) ID
Supplier: Thermo Scientific Chemicals
13471-RM
13471-RN
CAAA13471-RMEA
243.32
CAD
CAAA13471-RM
CAAA13471-RN
Copper ≥99.9% (metals basis), tubing 3.175 mm (0.125 in) OD, 1.5 mm (0.059 in) ID
Copper
MDL: MFCD00010965
Purity: 99.9%
CAS No.: 7440-50-8
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...