Sodium hydroxide, anhydrous ≥97.0% ACS low chloride

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Caustic soda

13455-22 13455-36 13455-A3 13455-A9 13455-F6
CAAA13455-22EA 68.73 CAD
CAAA13455-22 CAAA13455-36 CAAA13455-A3 CAAA13455-A9 CAAA13455-F6
Sodium hydroxide, anhydrous ≥97.0% ACS low chloride
Sodium hydroxide

In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal salts


Purity: 97.0% min CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Pellets Density: 2.13 pH: 14 Melting Point: 318° Boiling Point: 1390° Solubility: Soluble in water, alcohol and glycerol Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548


In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal salts

Formula: NaOH
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,8627

Order Now

Specification Test Results

Cl 0.005%
N 0.1%
PO9 0.001%
SO3 0.003%
Heavy Metals (as Ag) 0.002%
Fe 0.001%
Hg 0.1 ppm
Ca 0.005%
MG 0.002%
Ni 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR