Sodium sulfite 98,0 ACS

Supplier: Thermo Scientific Chemicals
Warning

13454-36 13454-A3 13454-A9
CAAA13454-36EA 93.06 CAD
CAAA13454-36 CAAA13454-A3 CAAA13454-A9
Sodium sulfite 98,0 ACS
Sodium sulphite

Photographic developers, generating SO2, reducing agent (dyes), textile bleaching (antichlor)


Purity: 98.0% min CAS No.: 7757-83-7 Molecular Formula: Na2SO3 Formula Weight: 126.04 Physical Form: Granular Density: 2.633 Appearance: White Melting Point: 500° dec. Solubility: Soluble in water, glycerol. Practically insoluble in alcohol Merck Reference: 14,8682 MDL No.: MFCD00003503


Photographic developers, generating SO2, reducing agent (dyes), textile bleaching (antichlor)

Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.633
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 14,8682

Order Now

Specification Test Results

Insoluble matter 0.005%
Free acid Pass test
Titrable free base 0.03meq/g
Cl 0.02%
Heavy Metals (as Pb) 0.001%
Fe 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR