Sodium chromate tetrahydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
13453-30
13453-A1
CAAA13453-30EA
125.45
CAD
CAAA13453-30
CAAA13453-A1
Sodium chromate tetrahydrate, REacton®, Reagent Grade
Sodium chromate tetrahydrate
Corrosion inhibitor, pigment manufacture
CAS No.: 10034-82-9
Molecular Formula: Na2CrO4.4H2O
Formula Weight: 234.04 (161.97anhy)
Physical Form: Crystalline
Density: 2.73
Sensitivity: Hygroscopic
Merck Reference: 14,8601
UN No.: 3288
MDL No.: MFCD00149167
Corrosion inhibitor, pigment manufacture
Formula:
CrNa₂O₄·4H₂O Melting Pt: 792.2 °C Density: 2.73 Storage Temperature: Ambient |
MDL Number:
MFCD00149167 CAS Number: 10034-82-9 EINECS: 231-889-5 UN: 3288 ADR: 6.1,II Merck Index: 14,8601 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...