di-Potassium oxalate monohydrate 98.5-101.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxalic acid potassium salt monohydrate, Potassium oxalate hydrate, Potassium oxalate monohydrate, Potassium ethanedioate monohydrate
MDL: MFCD00150033
Beilstein Registry No.: 3752576
Soluble in water
Purity: 98.8-101.0%
CAS No.: 6487-48-5
Molecular Formula: C2K2O4.H2O
Formula Weight: 184.23 (166.22anhy)
Physical Form: Granular/powder
Density: 2.13
Appearance: White
Odor: Odorless
pH: 6-8.5
Melting Point:
Formula:
(COOK)₂·H₂O MW: 184.23 g/mol Melting Pt: 356 Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00150033 CAS Number: 6487-48-5 EINECS: 209-506-8 UN: 2811 ADR: 6.1 III Merck Index: 14,7651 |
Specification Test Results
Insoluble matter | 0.01% |
Neutrality | Pass test |
Cl | 0.002% |
SO3 | 0.01% |
NH3 | 0.002% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.001% |
NA | 0.02% |
Substances darkened by sulfuric acid | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...