Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potash caustic, Caustic potash

13451-18 13451-36 13451-A3 13451-A9
CAAA13451-18EA 60.67 CAD
CAAA13451-18 CAAA13451-36 CAAA13451-A3 CAAA13451-A9
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
Potassium hydroxide

In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagent


Purity: 85% min CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Pellets Density: 2.044 pH: 13,5 Melting Point: 360° Boiling Point: 1320-1324° Solubility: Soluble in water, alcohol, and glycerol. Slightly soluble in ether Merck Reference: 14,7640 UN No.: 1813 MDL No.: MFCD00003553


In soap manufacture, in bleaching, as an electrolyte, as an absorbent, in dyestuffs, in liquid fertilizer, in electroplating, as a reagent

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320…1324 °C
Melting Pt: 360 °C
Density: 2.044
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8 II
REACH: 01-2119487136-33
Merck Index: 14,7640

Order Now

Specification Test Results

Cl 0.01%
Nitrogen compounds (as N) 0.1%
PO10 Pass test
SO3 0.003%
Heavy Metals (as Ag) 0.001%
Fe 0.001%
Ni 0.001%
Ca 0.005%
MG 0.002%
NA 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR