Potassium acetate ≥99% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
13449-22
13449-36
13449-A3
13449-F7
CAAA13449-22EA
52.14
CAD
CAAA13449-22
CAAA13449-36
CAAA13449-A3
CAAA13449-F7
Potassium acetate ≥99% ACS
Potassium acetate
MDL: MFCD00012458
Beilstein Registry No.: 3595449
Soluble in water and alcohol
Fieser: 1,906 6,475
Purity: 99% min
CAS No.: 127-08-2
Molecular Formula: C2H3KO2
Formula Weight: 98.15
Physical Form: Crystalline
Density: 1.57
Appearance: White
Melting Point: 292°
Solubility: Soluble in water and alcohol
Beilstein Reference: 3595449
Merck Reference: 14,7605
MDL No.: MFCD00012458
Formula:
CH₃COOK Melting Pt: 292 °C Density: 1.57 Flash Pt: >250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 EINECS: 204-822-2 Merck Index: 14,7605 |
|
Specification Test Results
Insoluble matter | 0.005% |
pH of a 5 % solution | 6.5-9.0 at 25° |
Cl | 0.003% |
PO3 | 0.00001 |
SO3 | 0.002% |
Ca | 0.005% |
MG | 0.002% |
Heavy metals (as Pb) | 5 ppm |
Fe | 5 ppm |
NA | 0.03% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...