Ammonium iron (II) sulfate hexahydrate 98.5-101.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
13448-36
13448-A3
CAAA13448-36EA
134.93
CAD
CAAA13448-36
CAAA13448-A3
Ammonium iron (II) sulfate hexahydrate 98.5-101.5% ACS
Ammonium iron (II) sulphate hexahydrate
Polymerization catalyst, photography
Purity: 98.5-101.5%
CAS No.: 7783-85-9
Molecular Formula: H8FeN2O8S2.6H2O
Formula Weight: 392.13 (284.04anhy)
Physical Form: Crystalline
Density: 1.864
Appearance: Pale bluish green
Odor: Odorless
Melting Point: ca 100° dec.
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,521
MDL No.: MFCD00150530
Polymerization catalyst, photography
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O Melting Pt: ∼ 100 °C (dec.) Density: 1.864 Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-382-4 Merck Index: 14,521 |
Specification Test Results
Insoluble matter | 0.01% |
PO3 | 0.003% |
Ca | 0.005% |
Cu | 0.003% |
MG | 0.002% |
Ferric ion (Fe??) | 0.01% |
Mn | 0.0001 |
K | 0.00002 |
NA | 0.0002 |
Zn | 0.003% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...