Lithium aluminum hydride ≥95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate

13442-14 13442-22 13442-36 13442-51 13442-59
CAAA13442-14EA 108.39 CAD
CAAA13442-14 CAAA13442-22 CAAA13442-36 CAAA13442-51 CAAA13442-59
Lithium aluminum hydride ≥95%
Lithium aluminum hydride

Reducing agent for pharmaceuticals, perfumes, and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizations


Purity: 95% min CAS No.: 16853-85-3 Molecular Formula: LiAlH4 Formula Weight: 37.95 Physical Form: Powder Density: 0.917 Appearance: White to grey Odor: Odorless Melting Point: 125° dec. Solubility: Decomposes in water. Soluble in ether Merck Reference: 14,349 UN No.: 1410 MDL No.: MFCD00011075


Reducing agent for pharmaceuticals, perfumes, and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizations

Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125 (dec.)
Density: 0.917
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3 I
Merck Index: 14,349

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR