Sodium phosphate, anhydrous, tech.

Supplier: Thermo Scientific Chemicals
Warning

13438-A1 13438-A7
CAAA13438-A1EA 98.43 CAD
CAAA13438-A1 CAAA13438-A7
Sodium phosphate, anhydrous, tech.
tri-Sodium phosphate

MDL: MFCD00003510


CAS No.: 7601-54-9 Molecular Formula: Na3O4P Formula Weight: 163.94 Physical Form: -100 Mesh Powder Sensitivity: Hygroscopic Appearance: White Merck Reference: 14,8662 UN No.: 3262 MDL No.: MFCD00003510

Formula: Na₃O₄P
Melting Pt: 73.3…76.7 °C
Density: 2.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003510
CAS Number: 7601-54-9
EINECS: 231-509-8
UN: 3262
ADR: 8,III
Merck Index: 14,8662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR