Disodium hydrogen phosphate, anhydrous ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic

13437-30 13437-A1 13437-A7
CAAA13437-30EA 69.99 CAD
CAAA13437-30 CAAA13437-A1 CAAA13437-A7
Disodium hydrogen phosphate, anhydrous ≥99.0% ACS
di-Sodium hydrogen phosphate

MDL: MFCD00003496 Notes: Suitable for buffer solutions


Purity: 99.0% min CAS No.: 7558-79-4 Molecular Formula: HNa2O4P Formula Weight: 141.98 Physical Form: Granular Sensitivity: Hygroscopic Merck Reference: 14,8659 MDL No.: MFCD00003496

Formula: Na₂HPO₄
Melting Pt: 250 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003496
CAS Number: 7558-79-4
EINECS: 231-448-7
REACH: 01-2119489797-11
Merck Index: 14,8659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR