Tetrapotassium diphosphate ≥95%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
13436-A1
13436-A7
CAAA13436-A1EA
87.85
CAD
CAAA13436-A1
CAAA13436-A7
Tetrapotassium diphosphate ≥95%, Technical Grade
tetra-Potassium diphosphate
MDL: MFCD00011393
Soluble in water
Purity: 95%
CAS No.: 7320-34-5
Molecular Formula: K4P2O7
Formula Weight: 330.34
Physical Form: Powder
Density: 2.33
Appearance: White
Melting Point: 1090°
Boiling Point: dec.
Solubility: Soluble in water
Merck Reference: 14,7663
MDL No.: MFCD00011393
Formula:
K₄O₇P₂ Boiling Pt: Dec. Melting Pt: 1090 °C Density: 2.33 Storage Temperature: Ambient |
MDL Number:
MFCD00011393 CAS Number: 7320-34-5 EINECS: 230-785-7 Merck Index: 14,7663 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...