Hafnium ≥99.7% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 1.57 mm (0.0618 in)

Supplier: Thermo Scientific Chemicals

13435-BU 13435-BY 13435-CC
CAAA13435-BUEA 95.91 CAD
CAAA13435-BU CAAA13435-BY CAAA13435-CC
Hafnium ≥99.7% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 1.57 mm (0.0618 in)
Hafnium

MDL: MFCD00011032


Purity: 99.7% CAS No.: 7440-58-6 Physical Form: ≈25.34g/m Odor: Odorless MDL No.: MFCD00011032

Formula: Hf
MW: 178.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS Number: 7440-58-6
EINECS: 231-166-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR