Sodium hydride 57 - 63% dispersion in mineral oil
Supplier: Thermo Scientific Chemicals
|
Danger
|
13431-18
13431-36
CAAA13431-18EA
71.89
CAD
CAAA13431-18
CAAA13431-36
Sodium hydride 57 - 63% dispersion in mineral oil
Sodium hydride
MDL: MFCD00003471
CAS No.: 7646-69-7
Molecular Formula: HNa
Formula Weight: 24.00
Physical Form: Moist powder
Density: 0.92
Sensitivity: Air and Moisture Sensitive
Appearance: Off-white to gray
Odor: Odorless
Merck Reference: 14,8625
UN No.: 1427
MDL No.: MFCD00003471
Formula:
HNa MW: 24 g/mol Density: 0.92 Storage Temperature: Ambient |
MDL Number:
MFCD00003471 CAS Number: 7646-69-7 EINECS: 231-587-3 UN: 1427 ADR: 4.3,I Merck Index: 14,8625 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...