Lithium molybdate(VI) 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Molybdic acid lithium salt, Lithium molybdenum oxide, Lithium molybdate, di-Lithium molybdate
13427-22
13427-36
CAAA13427-22EA
95.12
CAD
CAAA13427-22
CAAA13427-36
Lithium molybdate(VI) 99+%
Lithium molybdate(VI)
MDL: MFCD00016178
Purity: 99+%
CAS No.: 13568-40-6
Molecular Formula: Li2MoO4
Formula Weight: 173.83
Physical Form: Powder
Density: 2.66
Appearance: White
Odor: Odorless
Melting Point: 705°
MDL No.: MFCD00016178
Formula:
Li₂MoO₄ Melting Pt: 705 °C Density: 2,66 Storage Temperature: Ambient |
MDL Number:
MFCD00016178 CAS Number: 13568-40-6 EINECS: 236-977-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...