di-Lithium tetraborate 99 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium tetraborate, Dilithium tetraborate

13419-18 13419-30 13419-A1
CAAA13419-18EA 71.42 CAD
CAAA13419-18 CAAA13419-30 CAAA13419-A1
di-Lithium tetraborate 99 Metals basis
di-Lithium tetraborate

MDL: MFCD00011083


Purity: 99% CAS No.: 12007-60-2 Molecular Formula: B4Li2O7 Formula Weight: 169.12 Physical Form: Powder Density: 2.4 Appearance: White Odor: Odorless Melting Point: 930° Merck Reference: 14,5524 MDL No.: MFCD00011083

Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918
Density: 2.4
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR