Lithium carbonate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
13418-36
13418-A3
13418-A9
CAAA13418-36EA
249.64
CAD
CAAA13418-36
CAAA13418-A3
CAAA13418-A9
Lithium carbonate 99%
Lithium carbonate
MDL: MFCD00011084
Soluble in water and dilute acids. Insoluble in alcohol
Purity: 99%
CAS No.: 554-13-2
Molecular Formula: Li2CO3
Formula Weight: 73.90
Physical Form: Powder
Density: 2.11
Appearance: White
Odor: Odorless
pH: 10-11.5
Melting Point: 723°
Boiling Point: 1310°dec.
Solubility: Soluble in water and dilute acids. Insoluble in alcohol
Merck Reference: 14,5527
MDL No.: MFCD00011084
Formula:
Li₂CO₃ Boiling Pt: 1310 °C (dec.) Melting Pt: 723 °C Density: 2.11 Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 14,5527 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...