Lithium acetate dihydrate ≥99.9%, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
13417-30
13417-A1
CAAA13417-30EA
89.9
CAD
CAAA13417-30
CAAA13417-A1
Lithium acetate dihydrate ≥99.9%, REacton®, Reagent Grade
Lithium acetate dihydrate
MDL: MFCD00066949
Beilstein Registry No.: 3564320
Freely soluble in water and alcohol
CAS No.: 6108-17-4
Molecular Formula: LiOOCCH3.2H2O
Formula Weight: 102.01 (65.98anhy)
Physical Form: Crystals
Density: 1.300
Appearance: White
Melting Point: 54-57°
Solubility: Freely soluble in water and alcohol
Beilstein Reference: 3564320
Merck Reference: 14,5521
MDL No.: MFCD00066949
Formula:
H₃CCOOLi·2H₂O Melting Pt: 54…57 °C Density: 1.300 Storage Temperature: Ambient |
MDL Number:
MFCD00066949 CAS Number: 6108-17-4 EINECS: 208-914-3 Merck Index: 14,5521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...