Lithium aluminate

Supplier: Thermo Scientific Chemicals

13412-18 13412-30 13412-A1
CAAA13412-18EA 178.54 CAD
CAAA13412-18 CAAA13412-30 CAAA13412-A1
Lithium aluminate
Lithium aluminate

MDL: MFCD00016172


CAS No.: 12003-67-7 Molecular Formula: LiAlO2 Formula Weight: 65.92 Physical Form: Powder Density: 2.62 Odor: Odorless MDL No.: MFCD00016172

Formula: AlLiO₂
Melting Pt: >1625 °C
Density: 2.62
Storage Temperature: Ambient
MDL Number: MFCD00016172
CAS Number: 12003-67-7
EINECS: 234-434-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR