Lithium titanate
Supplier: Thermo Scientific Chemicals
13411-18
13411-30
CAAA13411-18EA
138.57
CAD
CAAA13411-18
CAAA13411-30
Lithium titanate
Lithium titanate
MDL: MFCD00016181
CAS No.: 12031-82-2
Molecular Formula: Li2TiO3
Formula Weight: 109.78
Physical Form: Powder
Odor: Odorless
Melting Point: 1520-1564°
MDL No.: MFCD00016181
Formula:
Li₂O₃Ti Melting Pt: 1520…1564 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016181 CAS Number: 12031-82-2 EINECS: 234-759-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...