Lithium bromide, anhydrous ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
Purity: 99% min
CAS No.: 7550-35-8
Molecular Formula: BrLi
Formula Weight: 86.85
Physical Form: Crystalline
Density: 3.464
Appearance: White
Odor: Odorless
Melting Point: 550°
Boiling Point: 1265°
Solubility: Very soluble in water and alcohol. Soluble in methanol, acetone, and glycol. Slightly soluble in pyridine
Merck Reference: 14,5526
MDL No.: MFCD00011077
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
Formula:
LiBr MW: 86.84 g/mol Boiling Pt: 1265 Melting Pt: 550 Density: 3.464 Storage Temperature: Ambient |
MDL Number:
MFCD00011077 CAS Number: 7550-35-8 EINECS: 231-439-8 Merck Index: 14,5526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...