Lithium hydroxide, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13407-22
013407-14
013407-36
CAAA13407-22EA
104.44
CAD
CAAA13407-22
AA13407-14
AAA13407-36
Lithium hydroxide, anhydrous 98%
Lithium hydroxide
Anhydrous
Purity: 98%
CAS No.: 1310-65-2
Molecular Formula: LiOH
Formula Weight: 23.95
Physical Form: -4+14 Mesh
Density: 2.54
Appearance: White
Odor: Odorless
pH: 14
Melting Point: 462°
Boiling Point: 924°
Solubility: Soluble in water. Slightly soluble in alcohol
Merck Reference: 14,5534
UN No.: 2680
MDL No.: MFCD00011095
Formula:
LiOH Boiling Pt: 924 °C Melting Pt: 462 °C Density: 2.54 Storage Temperature: Ambient |
MDL Number:
MFCD00011095 CAS Number: 1310-65-2 EINECS: 215-183-4 UN: 2680 ADR: 8,II Merck Index: 14,5534 |
Specification Test Results
Form | -4+14 Mesh |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...